MilliporeSigma
  • Dilute acid pretreatment of lignocellulose for whole slurry ethanol fermentation.
Dilute acid pretreatment of lignocellulose for whole slurry ethanol fermentation.
Bioresource technology (2013-02-12)
Young Hoon Jung, In Jung Kim, Hyun Kyung Kim, Kyoung Heon Kim
ABSTRACT
Dilute sulfuric acid pretreatment of oil palm empty fruit bunches (EFB) followed by the whole slurry fermentation of the pretreated EFB slurry was investigated. The optimized pretreatment conditions were at 1% (w/v) sulfuric acid with 3 min ramping to 190 °C in a microwave digester. Pretreated and washed EFB exhibited enzymatic digestibility of 88.5% of theoretical glucose yield after 48 h of hydrolysis. When the whole slurry of pretreated and neutralized EFB was used in simultaneous saccharification and fermentation (SSF) using cellulase and Saccharomyces cerevisiae, sulfuric acid-pretreated EFB resulted in 52.5% of theoretical ethanol yield based on total glucan in the untreated initial EFB after 72 h of SSF. When pretreated EFB slurry was treated with activated carbon before subjecting to SSF, the SSF furnished 87.5% ethanol yield based on the initial glucan content in untreated EFB (after 48 h of SSF).
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC