MilliporeSigma
Novel sulfamides as potential carbonic anhydrase isoenzymes inhibitors.
Bioorganic & medicinal chemistry (2013-02-12)
Akın Akıncıoğlu, Yusuf Akbaba, Hülya Göçer, Süleyman Göksu, İlhami Gülçin, Claudiu T Supuran
ABSTRACT
Sulfamides represent an important class of biologically active compounds. A series of novel sulfamides were synthesized from 1-aminoindanes, 1-aminotetralin, 2-aminoindanes and 2-aminotetralin via the reactions of free amines, benzyl alcohol and chlorosulfonyl isocyanate (CSI) followed by hydrogenolysis of the obtained sulfamoylcarbamates. Carbonic anhydrase (CA, EC 4.2.1.1) inhibitory effects of the new sulfamides have been investigated. The human (h) isozymes hCA I and hCA II have been investigated in this study by using an esterase assay with 4-nitrophenyl acetate as substrate. The new sulfamides showed inhibition constants in the micro-submicromolar range, with one compound (N-(indane-1-yl)sulfamide) showing a Ki of 0.45μM against hCA I and of 1.07μM against hCA II.
Sigma-Aldrich
4-Nitrophenyl acetate, esterase substrate