MilliporeSigma
Total synthesis of kopsinine.
Organic letters (2013-02-09)
Jian Xie, Amanda L Wolfe, Dale L Boger
ABSTRACT
The use of a powerful intramolecular [4 + 2]/[3 + 2] cycloaddition cascade of an 1,3,4-oxadiazole in the divergent total synthesis of kopsinine (1), featuring an additional unique SmI(2)-promoted transannular cyclization reaction for formation of the bicyclo[2.2.2]octane central to its hexacyclic ring system, is detailed.
Sigma-Aldrich
Octane, reagent grade, 98%
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Sigma-Aldrich
Octane, anhydrous, ≥99%
Supelco
Octane, analytical standard