MilliporeSigma
Blood optical clearing studied by optical coherence tomography.
Journal of biomedical optics (2013-02-08)
Olga Zhernovaya, Valery V Tuchin, Martin J Leahy
ABSTRACT
The main limitation of optical imaging techniques for studying biological tissues is light scattering leading to decreasing of transmittance, which lowers the imaging quality. In this case, an immersion method for optical clearing of biological tissues can provide a possible solution to this problem, because the application of biocompatible clearing agents can reduce light scattering. Optical clearing represents a promising approach to increasing the imaging depth for various techniques, for example, various spectroscopy and fluorescent methods, and optical coherence tomography (OCT). We investigate the improvement of light penetration depth in blood after application of polyethylene glycol, polypropylene glycol, propylene glycol, and hemoglobin solutions using an OCT system. Influence of clearing agents on light transport in tissues and blood was also investigated in the mouse tail vein.
Sigma-Aldrich
Poly(propylene glycol), average Mn ~425
Sigma-Aldrich
Poly(propylene glycol), average Mn ~4,000
Sigma-Aldrich
Poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Poly(propylene glycol), average Mn ~2,700
Sigma-Aldrich
Poly(propylene glycol), average Mn ~1,000
Sigma-Aldrich
Poly(propylene glycol), average Mn ~725
Sigma-Aldrich
Polypropylene glycol, P 2,000
Sigma-Aldrich
Polypropylene glycol, P 400
Sigma-Aldrich
(R)-(−)-1,2-Propanediol, 96%
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
(S)-(+)-1,2-Propanediol, 96%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%