MilliporeSigma
  • DNA binding, nuclease activity, DNA photocleavage and cytotoxic properties of Cu(II) complexes of N-substituted sulfonamides.
DNA binding, nuclease activity, DNA photocleavage and cytotoxic properties of Cu(II) complexes of N-substituted sulfonamides.
Journal of inorganic biochemistry (2013-02-07)
José Luis García-Giménez, Javier Hernández-Gil, Aloma Martínez-Ruíz, Alfonso Castiñeiras, Malva Liu-González, Federico V Pallardó, Joaquín Borrás, Gloria Alzuet Piña
ABSTRACT
Ternary copper(II) complexes [Cu(NST)2(phen)] (1) and [Cu(NST)2(NH3)2]·H2O (2) [HNST=N-(4,5-dimethylthiazol-2-yl)naphthalene-1-sulfonamide] were prepared and characterized by physico-chemical techniques. Both 1 and 2 were structurally characterized by X-ray crystallography. The crystal structures show the presence of a distorted square planar CuN4 geometry in which the deprotonated sulfonamide, acting as monodentate ligand, binds to the metal ion through the thiazole N atom. Both complexes present intermolecular π-π stacking interactions between phenanthroline rings (compound 1) and between naphthalene rings (compound 2). The interaction of the complexes with CT DNA was studied by means of thermal denaturation, viscosity measurements and fluorescence spectroscopy. The complexes display good binding propensity to the calf thymus DNA giving the order: 1>2. Complex 1, which has a higher capability for binding to DNA, showed better nuclease activity than 2 in the presence of ascorbate/H2O2. Both the kinetics and the mechanism of the DNA cleavage reaction were investigated. Furthermore, complex 1 showed efficient photo-induced DNA cleavage activity on irradiation with UV light in the absence of any external reagent. The UV light induced DNA cleavage follows a photo-redox pathway with generation of hydroxyl radicals as reactive species. In addition, the cytotoxic properties of both complexes (1 and 2) were evaluated in human cancer cells (HeLa, Caco-2 and MDA-468). The low IC50 values, in particular those against Caco-2, have indicated that the compounds can be considered as promising chemotherapeutic agents.
Sigma-Aldrich
Deoxyribonucleic acid from calf thymus, Type XV, Activated, lyophilized powder
Sigma-Aldrich
Deoxyribonucleic acid from calf thymus, Genomic, unsheared
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from calf thymus, lyophilized powder