MilliporeSigma
  • Diazinon residues and degradation kinetics for grapes under field conditions.
Diazinon residues and degradation kinetics for grapes under field conditions.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2013-02-05)
Ehssan Torabi, Khalil Talebi
ABSTRACT
The dissipation of diazinon (O,O-diethyl O-2-isopropyl-6-methylpyrimidin-4-yl phosphorothioate) in grapes was investigated to determine its pre-harvest interval (PHI). Diazinon was applied to grapes at the recommended dosage (0.9 g a.i. L(-1)) and twice the recommended dosage (1.8 g a.i. L(-1)) three times, at the fruit formation stage, the sour stage, and the ripening stage, in a field trial with three replications. Samples were taken at 0, 1, 2, 4, 7, 11, 15, 20, 26 and 32 days after spraying. The residue was extracted using a water/methanol/acetonitrile (1:1:1, v/v/v) solvent and solid phase extraction was employed for cleanup. Quantitative analysis was performed using a gas chromatograph equipped with a nitrogen-phosphorus detector. The dissipation trend for the recommended dosage and twice the recommend dosage followed the simple first-order kinetic model (SFOK) (DT(50) = 3.29 days, DT(90) = 11 days, PHI = 13.5 days) and first-order double-exponential decay (FODED) model (DT(50) = 1.08 days, DT(90) = 5.82 days, PHI = 15.29 days), respectively. The average initial deposit of diazinon at the recommended dosage was 9.04 mg kg(-1)and for twice the recommended dosage was 27.38 mg kg(-1)and it dissipated rapidly within days of spraying.
Product Description
Supelco
Diazinon, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Diazinon, PESTANAL®, analytical standard
Supelco
Diazinon solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard