MilliporeSigma
  • Potassium and ionic strength effects on the conformational and thermal stability of two aldehyde dehydrogenases reveal structural and functional roles of K⁺-binding sites.
Potassium and ionic strength effects on the conformational and thermal stability of two aldehyde dehydrogenases reveal structural and functional roles of K⁺-binding sites.
PloS one (2013-02-01)
Georgina Garza-Ramos, Carlos Mújica-Jiménez, Rosario A Muñoz-Clares
ABSTRACT
Many aldehyde dehydrogenases (ALDHs) have potential potassium-binding sites of as yet unknown structural or functional roles. To explore possible K(+)-specific effects, we performed comparative structural studies on the tetrameric betaine aldehyde dehydrogenase from Pseudomonas aeruginosa (PaBADH) and on the dimeric BADH from spinach (SoBADH), whose activities are K(+)-dependent and K(+)-independent, respectively, although both enzymes contain potassium-binding sites. Size exclusion chromatography, dynamic light scattering, far- and near-UV circular dichroism, and extrinsic fluorescence results indicated that in the absence of K(+) ions and at very low ionic strength, PaBADH remained tetrameric but its tertiary structure was significantly altered, accounting for its inactivation, whereas SoBADH formed tetramers that maintained the native tertiary structure. The recovery of PaBADH native tertiary-structure was hyperbolically dependent on KCl concentration, indicating potassium-specific structuring effects probably arising from binding to a central-cavity site present in PaBADH but not in SoBADH. K(+) ions stabilized the native structure of both enzymes against thermal denaturation more than did tetraethylammonium (TEA(+)) ions. This indicated specific effects of potassium on both enzymes, particularly on PaBADH whose apparent T(m) values showed hyperbolical dependence on potassium concentration, similar to that observed with the tertiary structure changes. Interestingly, we also found that thermal denaturation of both enzymes performed in low ionic-strength buffers led to formation of heat-resistant, inactive soluble aggregates that retain 80% secondary structure, have increased β-sheet content and bind thioflavin T. These structured aggregates underwent further thermal-induced aggregation and precipitation when the concentrations of KCl or TEACl were raised. Given that PaBADH and SoBADH belong to different ALDH families and differ not only in amino acid composition but also in association state and surface electrostatic potential, the formation of this kind of β-sheet pre-fibrillar aggregates, not described before for any ALDH enzyme, appear to be a property of the ALDH fold.
Sigma-Aldrich
Tetraethylammonium chloride hydrate
Sigma-Aldrich
Tetraethylammonium iodide, 98%
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
Tetraethylammonium hydroxide solution, ~25% in methanol (~1.5 M)
Sigma-Aldrich
Tetraethylammonium chloride, ≥98% (titration)
Sigma-Aldrich
Tetraethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Tetraethylammonium chloride, for electrochemical analysis, ≥99.0%
Sigma-Aldrich
Tetraethylammonium bromide, ReagentPlus®, 99%
Supelco
Tetraethylammonium hydroxide solution, ~1.0 M (CH3CH2)4NOH in H2O, electrochemical grade
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 35 wt. % in H2O
Sigma-Aldrich
Tetraethylammonium bromide, reagent grade, 98%
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 20 wt. % in H2O