MilliporeSigma
  • Calcium, calpain, and calcineurin in low-frequency depression of transmitter release.
Calcium, calpain, and calcineurin in low-frequency depression of transmitter release.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-02-01)
Lorelei B Silverman-Gavrila, Moshe Praver, Donald L Mykles, Milton P Charlton
ABSTRACT
Low-frequency depression (LFD) of transmitter release occurs at phasic synapses with stimulation at 0.2 Hz in both isolated crayfish (Procambarus clarkii) neuromuscular junction (NMJ) preparations and in intact animals. LFD is regulated by presynaptic activity of the Ca(2+)-dependent phosphatase calcineurin (Silverman-Gavrila and Charlton, 2009). Since the fast Ca(2+) chelator BAPTA-AM inhibits LFD but the slow chelator EGTA-AM does not, the Ca(2+) sensor for LFD may be close to a Ca(2+) source at active zones. Calcineurin can be activated by the Ca(2+)-activated protease calpain, and immunostaining showed that both proteins are present at nerve terminals. Three calpain inhibitors, calpain inhibitor I, MDL-28170, and PD150606, but not the control compound PD145305, inhibit LFD both in the intact animal as shown by electromyograms and by intracellular recordings at neuromuscular junctions. Analysis of mini-EPSPs indicated that these inhibitors had minimal postsynaptic effects. Proteolytic activity in CNS extract, detected by a fluorescent calpain substrate, was modulated by Ca(2+) and calpain inhibitors. Western blot analysis of CNS extract showed that proteolysis of calcineurin to a fragment consistent with the constitutively active form required Ca(2+) and was blocked by calpain inhibitors. Inhibition of LFD by calpain inhibition blocks the reduction in phosphoactin and the depolymerization of tubulin that normally occurs in LFD, probably by blocking the dephosphorylation of cytoskeletal proteins by calcineurin. In contrast, high-frequency depression does not involve protein phosphorylation- or calpain-dependent mechanisms. LFD may involve a specific pathway in which local Ca(2+) signaling activates presynaptic calpain and calcineurin at active zones and causes changes of tubulin cytoskeleton.
Sigma-Aldrich
MDL 28170, ≥90% (TLC)