MilliporeSigma
  • [State of the dopamine system activity, as one of factors for the development of arterial hypertension and obesity].
[State of the dopamine system activity, as one of factors for the development of arterial hypertension and obesity].
Likars'ka sprava (2013-01-30)
V H Lyzogub, O V Dolynna, T V Bogdan, V O Sobol'
ABSTRACT
The discovery of 5 subtypes of dopamine receptors revealed their important role in development of arterial hypertension and obesity. Reduce of their functional activity or number with age conduces to the increase of tone of the sympathetic nervous system, dyspoiesis of leptin, hyperphagia, development of obesity and arterial hypertension. Prescription of dopamine agonists conduces to reduction of in, normalization of leptin, diminishing of appetite and body mass, decrease of blood pressure. Combination of arterial hypertension and obesity is recommended to determine dopamine excretion and to appoint dopamine agonists at level of < 600 nmol/24 hours.