MilliporeSigma
  • In vitro toxicological characterisation of the S-containing arsenic metabolites thio-dimethylarsinic acid and dimethylarsinic glutathione.
In vitro toxicological characterisation of the S-containing arsenic metabolites thio-dimethylarsinic acid and dimethylarsinic glutathione.
Toxicology (2013-01-29)
Larissa Leffers, Marlies Unterberg, Marc Bartel, Cornelia Hoppe, Imke Pieper, Julia Stertmann, Franziska Ebert, Hans-Ulrich Humpf, Tanja Schwerdtle
ABSTRACT
Inorganic arsenic is a well-documented, exposure relevant human carcinogen. A promising starting point to further understand the mechanisms behind inorganic arsenic carcinogenicity might be a formation of reactive, highly toxic metabolites during human arsenic metabolism. This study characterises the toxicity of recently identified S-containing arsenic metabolites in cultured human A549 lung adenocarcinoma epithelium cells. In direct comparison to arsenite, thio-dimethylarsinic acid (thio-DMA(V)) and dimethylarsinic glutathione (DMAG) exerted a 5- to 20-fold stronger cytotoxicity and showed a 2- to 20-fold higher cellular bioavailability, respectively. All three arsenicals disturbed cell cycle progression at cytotoxic concentrations, but failed to increase the level of reactive oxygen and nitrogen species (RONS) in healthy A549 cells. However, a strong disturbance of the oxidative defense system was observed after incubation with absolutely sub-cytotoxic, pico- to nanomolar concentrations of arsenite and thio-DMA(V), respectively. Thus, both GSH and GSSG levels were significantly decreased by up to 40%. Accordingly, RONS levels of oxidatively (H2O2) stressed cells were strongly increased by the arsenicals. Since in vivo RONS are permanently endogenously and exogenously produced, this boost of the existing oxidative stress by arsenite and thio-DMA(V) might contribute to the process of inorganic arsenic induced carcinogenicity.
Sigma-Aldrich
Cacodylic acid, ≥98%
Sigma-Aldrich
Cacodylic acid, ≥99.0%