MilliporeSigma
  • Choline acetate enhanced the catalytic performance of Candida rogusa lipase in AOT reverse micelles.
Choline acetate enhanced the catalytic performance of Candida rogusa lipase in AOT reverse micelles.
Colloids and surfaces. B, Biointerfaces (2013-01-29)
Luyan Xue, Yin Zhao, Lijie Yu, Yanwen Sun, Keqian Yan, Ying Li, Xirong Huang, Yinbo Qu
ABSTRACT
Choline acetate is an ionic liquid composed of a kosmotropic anion and a chaotropic cation. According to Hofmeister series, a kosmotropic anion and/or a chaotropic cation could stabilize an enzyme, thereby facilitating the retention of the catalytic activity of the enzyme. In this work, we first report the influence of choline acetate on the activity and stability of lipase in AOT/water/isooctane reverse micelles. The indicator reaction is the lipase-catalyzed hydrolysis of 4-nitrophenyl butyrate. The results show that a low level of choline acetate does not affect the microstructure of the AOT reverse micelles, but the ionic liquid can improve the catalytic efficiency of lipase. Fluorescence spectra show that a high level of choline acetate has an impact on the conformation of lipase, so the activation is mainly due to the influence of choline acetate on the nucleophilicity of water. Infrared spectra demonstrate that choline acetate can form stronger hydrogen bonds with water surrounding lipase, and therefore enhance the nucleophilicity of the water, which makes it easier to attack the acyl enzyme intermediate, thereby increasing the activity of the lipase-catalyzed hydrolysis of the ester. A study on the stability of lipase in AOT reverse micelles indicates that the ionic liquid is able to maintain the activity of lipase to a certain extent. The effect of choline acetate is consistent with that predicted based on Hofmeister series.
Sigma-Aldrich
2,2,4-Trimethylpentane, anhydrous, 99.8%
Supelco
2,2,4-Trimethylpentane, analytical standard
Sigma-Aldrich
2,2,4-Trimethylpentane, ACS reagent, ≥99.0%
Sigma-Aldrich
2,2,4-Trimethylpentane, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
2,2,4-Trimethylpentane, suitable for HPLC, ≥99%
Sigma-Aldrich
2,2,4-Trimethylpentane, ReagentPlus®, ≥99%
Sigma-Aldrich
2,2,4-Trimethylpentane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.5%
Supelco
Density Standard 692 kg/m3, H&D Fitzgerald Ltd. Quality