MilliporeSigma
  • The regio- and stereospecific intermolecular dehydrative alkoxylation of allylic alcohols catalyzed by a gold(I) N-heterocyclic carbene complex.
The regio- and stereospecific intermolecular dehydrative alkoxylation of allylic alcohols catalyzed by a gold(I) N-heterocyclic carbene complex.
Chemistry (Weinheim an der Bergstrasse, Germany) (2013-01-26)
Paramita Mukherjee, Ross A Widenhoefer
ABSTRACT
A 1:1 mixture of [AuCl(IPr)] (IPr = 1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidine) and AgClO(4) catalyzes the intermolecular dehydrative alkoxylation of primary and secondary allylic alcohols with aliphatic primary and secondary alcohols to form allylic ethers. These transformations are regio- and stereospecific with preferential addition of the alcohol nucleophile at the γ-position of the allylic alcohol syn to the departing hydroxyl group and with predominant formation of the E stereoisomer. The minor α regioisomer is formed predominantly through a secondary reaction manifold involving regioselective γ-alkoxylation of the initially formed allylic ether rather than by the direct α-alkoxylation of the allylic alcohol.
Sigma-Aldrich
Allyl alcohol, ≥99%
Sigma-Aldrich
Allyl alcohol, ≥98.5%