MilliporeSigma
A backbone lever-arm effect enhances polymer mechanochemistry.
Nature chemistry (2013-01-25)
Hope M Klukovich, Tatiana B Kouznetsova, Zachary S Kean, Jeremy M Lenhardt, Stephen L Craig
ABSTRACT
Mechanical forces along a polymer backbone can be used to bring about remarkable reactivity in embedded mechanically active functional groups, but little attention has been paid to how a given polymer backbone delivers that force to the reactant. Here, single-molecule force spectroscopy was used to directly quantify and compare the forces associated with the ring opening of gem-dibromo and gem-dichlorocyclopropanes affixed along the backbone of cis-polynorbornene and cis-polybutadiene. The critical force for isomerization drops by about one-third in the polynorbornene scaffold relative to polybutadiene. The root of the effect lies in more efficient chemomechanical coupling through the polynorbornene backbone, which acts as a phenomenological lever with greater mechanical advantage than polybutadiene. The experimental results are supported computationally and provide the foundation for a new strategy by which to engineer mechanochemical reactivity.
Sigma-Aldrich
Polybutadiene, cis, average Mw 200,000-300,000
Sigma-Aldrich
Polybutadiene, predominantly 1,2-addition, approx. 90% 1,2-vinyl
Sigma-Aldrich
Polybutadiene, average Mn ~5,000