MilliporeSigma
Embelin accelerates cutaneous wound healing in diabetic rats.
Journal of Asian natural products research (2013-01-19)
Pradeep T Deshmukh, Vipin B Gupta
ABSTRACT
This study reports the effect of embelin (1) on cutaneous wound in streptozotocin (STZ)-induced diabetic rats. The effect was studied using excision, incision, and dead space models. In diabetic rats, topical application of embelin 5% (w/w) ointment showed a significant increase in wound contraction and better epithelialization, thereby facilitating the healing. Embelin was also active by the oral route (25 and 50 mg/kg) in the incision and dead space wound models. In incision wound model, wound granulation tissues were removed on 8th post-wounding day, and the hydroxyproline, hexosamine, total protein, and DNA contents were determined. In STZ diabetic rats, topical and oral applications of embelin showed an increase in hydroxyproline, hexosamine, total protein, and DNA contents. It also showed a significant increase in wound breaking strength. Embelin significantly increased granuloma tissue weight and breaking strength in dead space model. These results indicated that embelin accelerated wound healing in diabetic rat.
Sigma-Aldrich
Embelin, ≥98% (HPLC), powder