MilliporeSigma
  • Dissociative adsorption of 3-chloropropyne on Si(111)-(7 × 7): binding and structure.
Dissociative adsorption of 3-chloropropyne on Si(111)-(7 × 7): binding and structure.
Langmuir : the ACS journal of surfaces and colloids (2013-01-19)
Yongping Zhang, Zhiqian Chen, Yuhong Man, Peitao Guo
ABSTRACT
To achieve silicon functionalization for the development of hybrid devices, multifunctional molecules may be employed to attach to the silicon surfaces. It is important to get a fundamental understanding about the molecule/silicon interface chemistry and the binding configuration. The surface chemistry of 3-chloropropyne (HC≡C-CH(2)Cl) on the Si(111)-(7 × 7) surface, as a model system for understanding the interaction of the multifunctional molecules with a silicon surface, was studied by X-ray photoelectron spectroscopy (XPS), high-resolution electron energy loss spectroscopy (HREELS), and density functional theory (DFT). The 3-chloropropyne adsorbs molecularly on the silicon surface at 110 K. A chemical reaction clearly occurs such that 3-choloropropyne bonds onto the Si(111)-(7 × 7) surface at room temperature by forming C-Si linkage through the cleavage of C-Cl bond, and preserving the ethyne C≡C triple bond. This functionalized silicon surface may act as an intermediate for the growth of multiple organic layers by further attaching other functional molecules.