MilliporeSigma
  • History of the discovery of the antipsychotic dopamine D2 receptor: a basis for the dopamine hypothesis of schizophrenia.
History of the discovery of the antipsychotic dopamine D2 receptor: a basis for the dopamine hypothesis of schizophrenia.
Journal of the history of the neurosciences (2013-01-18)
Bertha K Madras
ABSTRACT
The 1975 publication of Seeman et al. (Proc Nat Acad Sci, USA), reporting the discovery of the antipsychotic receptor in the brain, is a classic example of translational medicine research. In searching for a pathophysiological mechanism of psychosis, the team sought to identify sites that bound the antipsychotic drug haloperidol. Their criterion was that haloperidol bound to the site at one to two nanomoles per liter, corresponding to haloperidol concentrations found in spinal fluid or plasma water in treated patients. They requested de novo synthesis of tritiated haloperidol, and it readily detected specific haloperidol binding sites in brain striatum. With dopamine binding the haloperidol-labeled sites with higher potency than other neurotransmitters, the sites were named antipsychotic/dopamine receptors (now designated dopamine D2 receptors). Most significantly, they found that all antipsychotics bound these sites at concentrations and with a rank order of potencies that were directly related to the mean daily antipsychotic dose taken by patients with schizophrenia. Their findings enabled screening for new antipsychotics, initiated D2 receptor measurements in brain of living patients, and determination of minimum occupancy (65%) of D2 receptors for antipsychotic benefit. The collective work is generally viewed as providing a fundamental basis for the dopamine hypothesis of schizophrenia.
Product Description
Haloperidol, European Pharmacopoeia (EP) Reference Standard
Supelco
Haloperidol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Haloperidol, powder