MilliporeSigma
A novel breath test to directly measure use of vaginal gel and condoms.
AIDS and behavior (2013-01-17)
Ariane van der Straten, Helen Cheng, Scott Wasdo, Liz Montgomery, Karen Smith-McCune, Matthew Booth, Daniel Gonzalez, Hartmut Derendorf, Timothy E Morey, Donn M Dennis
ABSTRACT
We assessed the feasibility of a breath test to detect women's single or concurrent use of vaginal products by adding ester taggants to vaginal gel and condom lubricant. Healthy non-pregnant women were enrolled into a two-day cohort (N = 13) and a single-day cohort (N = 12) in San Francisco. Within each cohort, women were randomized (5:1) to tagged or untagged products, and inserted in a clinical setting: 4 mL of tenofovir placebo gel (ten tagged with 15 mg 2-pentyl acetate; three untagged), and an artificial phallus with a lubricated condom (11 tagged with 15 mg 2-butyl acetate; two untagged), on two separate days (two-day cohort) or concurrently (single-day cohort). Using a portable mini-gas chromatograph, the presence/absence of taggants was determined in breath specimens collected prior to, and at timed intervals following product exposure. Demographic, clinical and product use experience data were collected by structured interview. All participants completed all visits and inserted their assigned products. At 5 min post-insertion, the breath test was 100% accurate in identifying insertion of the tagged (or untagged) gel and/or condom. The half-life in breath of the two esters tested was <1 h with large variability between individuals, taggants and cohorts. Overall, among those receiving tagged product, six mild and two moderate product-related AEs were reported. All were transient and resolved spontaneously. Additional sensations included taste in mouth (N = 4) and scent (N = 5). The tagged products were well tolerated. This breath test has the potential to accurately and objectively monitor adherence to vaginal gel and condom used separately or concurrently.
Product Description
Supelco
Butyl acetate, analytical standard
Sigma-Aldrich
Butyl acetate, natural, ≥98%, FG
Sigma-Aldrich
Amyl acetate, ≥99%, FG
Sigma-Aldrich
Butyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Butyl acetate, anhydrous, ≥99%
Sigma-Aldrich
Butyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Pentyl acetate, 99%
Sigma-Aldrich
Pentyl acetate, puriss. p.a., ≥98.5% (GC)
Supelco
Butyl acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butyl acetate, ReagentPlus®, 99.5%
Sigma-Aldrich
Butyl acetate, suitable for HPLC, 99.7%