MilliporeSigma
  • Palladium catalyzed cyclizations of oxime esters with 1,1-disubstituted alkenes: synthesis of α,α-disubstituted dihydropyrroles and studies towards an asymmetric protocol.
Palladium catalyzed cyclizations of oxime esters with 1,1-disubstituted alkenes: synthesis of α,α-disubstituted dihydropyrroles and studies towards an asymmetric protocol.
Chemical communications (Cambridge, England) (2013-01-17)
Adele Faulkner, James S Scott, John F Bower
ABSTRACT
We report efficient Pd-catalyzed cyclizations of oxime esters with 1,1-disubstituted alkenes as the basis of a general entry to α,α-disubstituted pyrrolidine derivatives. We also demonstrate that catalytic asymmetric variants of this chemistry are feasible by employing a suitable chiral ligand.
Sigma-Aldrich
Pyrrolidine, FG
Sigma-Aldrich
Pyrrolidine, ≥99.0%
Sigma-Aldrich
Pyrrolidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Pyrrolidine, 99%