MilliporeSigma
[Preparation and pharmacokinetics of brucine hydrogel patch].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2013-01-17)
Jun Chen, Lei Li, Yan Qi, Zhi-Peng Chen, Bao-Chang Cai
ABSTRACT
To investigate the effect of dose on pharmacokinetic properties of brucine hydrogel patch. The plasma concentration of brucine was determined by HPLC. Brucine hydrogel patch was prepared and its pharmaceutical characterization was investigated. After transdermal administration of different dose brucine hydrogel patch; Plasma concentration versus time profiles were determined and pharmacokinetic parameters were calculated by DAS program. The pharmaceutical properties of brucine hydrogel patch were satisfactory. The AUC0-1 values were 7.24 +/- 0.61, 16.02 +/- 2.34 and 54.84 +/- 26.59 microg x h/mL after administration of 30, 60 and 180 mg/kg brucine hydrogel patch, respectively. The corresponding C(max) values were 0.73 +/- 0.23, 1.45 +/- 0.28 and 4.59 +/- 1.85 microg/mL, respectively. And the corresponding T(max) values were 8.67 +/- 2.07, 11.67 +/- 2.66 and 8.33 +/- 2.65 h, respectively. The pharmacokinetic properties of brucine do not vary with the dose of brucine hydrogel patch.
Sigma-Aldrich
Carboxymethylcellulose sodium salt, medium viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, ultra high viscosity, highly purified
Sigma-Aldrich
Carboxymethylcellulose sodium salt, High viscosity
Sigma-Aldrich
Carboxymethylcellulose sodium salt, low viscosity
Sigma-Aldrich
Strychnine
Sigma-Aldrich
Brucine, anhydrous, 98%
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.9
Sigma-Aldrich
Sodium carboxymethyl cellulose, viscosity 50-200 cP , c=4% H2O at 25­°C
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 1.2
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~700,000
Sigma-Aldrich
Sodium carboxymethyl cellulose, average Mw ~250,000, degree of substitution 0.7
Sigma-Aldrich
Carboxymethylcellulose sodium, meets USP testing specifications, Medium viscosity