MilliporeSigma
Antitumor activity of a pyrrole-imidazole polyamide.
Proceedings of the National Academy of Sciences of the United States of America (2013-01-16)
Fei Yang, Nicholas G Nickols, Benjamin C Li, Georgi K Marinov, Jonathan W Said, Peter B Dervan
ABSTRACT
Many cancer therapeutics target DNA and exert cytotoxicity through the induction of DNA damage and inhibition of transcription. We report that a DNA minor groove binding hairpin pyrrole-imidazole (Py-Im) polyamide interferes with RNA polymerase II (RNAP2) activity in cell culture. Polyamide treatment activates p53 signaling in LNCaP prostate cancer cells without detectable DNA damage. Genome-wide mapping of RNAP2 binding shows reduction of occupancy, preferentially at transcription start sites, but occupancy at enhancer sites is unchanged. Polyamide treatment results in a time- and dose-dependent depletion of the RNAP2 large subunit RPB1 that is preventable with proteasome inhibition. This polyamide demonstrates antitumor activity in a prostate tumor xenograft model with limited host toxicity.
Product Description
Imidazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Imidazole, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole hydrochloride
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole sodium derivative, technical grade
Sigma-Aldrich
Imidazole, for molecular biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O