MilliporeSigma
Silicon carbide: a versatile material for biosensor applications.
Biomedical microdevices (2013-01-16)
Alexandra Oliveros, Anthony Guiseppi-Elie, Stephen E Saddow
ABSTRACT
Silicon carbide (SiC) has been around for more than 100 years as an industrial material and has found wide and varied applications because of its unique electrical and thermal properties. In recent years there has been increased attention to SiC as a viable material for biomedical applications. Of particular interest in this review is its potential for application as a biotransducer in biosensors. Among these applications are those where SiC is used as a substrate material, taking advantage of its surface chemical, tribological and electrical properties. In addition, its potential for integration as system on a chip and those applications where SiC is used as an active material make it a suitable substrate for micro-device fabrication. This review highlights the critical properties of SiC for application as a biosensor and reviews recent work reported on using SiC as an active or passive material in biotransducers and biosensors.
Sigma-Aldrich
Silicon carbide, nanopowder, <100 nm particle size
Sigma-Aldrich
Silicon carbide, -200 mesh particle size
Sigma-Aldrich
Silicon carbide, −400 mesh particle size, ≥97.5%