MilliporeSigma
  • Direct access to enantioenriched spiroacetals through asymmetric relay catalytic three-component reaction.
Direct access to enantioenriched spiroacetals through asymmetric relay catalytic three-component reaction.
Organic letters (2013-01-15)
Hua Wu, Yu-Ping He, Liu-Zhu Gong
ABSTRACT
The gold(I)/chiral Brønsted acid relay catalysis enabled a highly stereoselective three-component reaction of salicylaldehydes, anilines, and alkynols to give aromatic spiroacetals in high yields and stereoselectivities.
Product Description
Supelco
Salicylaldehyde, analytical standard
Sigma-Aldrich
Salicylaldehyde, redist., ≥99.0% (GC)
Sigma-Aldrich
Salicylaldehyde, ≥98%, FG
Sigma-Aldrich
Salicylaldehyde, reagent grade, 98%