MilliporeSigma
  • Promiscuous catalysis of asymmetric Michael-type additions of linear aldehydes to β-nitrostyrene by the proline-based enzyme 4-oxalocrotonate tautomerase.
Promiscuous catalysis of asymmetric Michael-type additions of linear aldehydes to β-nitrostyrene by the proline-based enzyme 4-oxalocrotonate tautomerase.
Chembiochem : a European journal of chemical biology (2013-01-11)
Yufeng Miao, Edzard M Geertsema, Pieter G Tepper, Ellen Zandvoort, Gerrit J Poelarends
ABSTRACT
Exploiting catalytic promiscuity: The proline-based enzyme 4-oxalocrotonate tautomerase (4-OT) promiscuously catalyzes asymmetric Michael-type additions of linear aldehydes--ranging from acetaldehyde to octanal--to trans-β-nitrostyrene in aqueous solvent. The presence of 1.4 mol% of 4-OT effected formation of the anticipated γ-nitroaldehydes in fair to good yields with dr values of up to 93:7 and ee values of up to 81 %.
Sigma-Aldrich
trans-β-Nitrostyrene, 99%