MilliporeSigma
  • Covalently cross-linked sulfone polybenzimidazole membranes with poly(vinylbenzyl chloride) for fuel cell applications.
Covalently cross-linked sulfone polybenzimidazole membranes with poly(vinylbenzyl chloride) for fuel cell applications.
ChemSusChem (2013-01-11)
Jingshuai Yang, David Aili, Qingfeng Li, Lars N Cleemann, Jens Oluf Jensen, Niels J Bjerrum, Ronghuan He
ABSTRACT
Covalently cross-linked polymer membranes were fabricated from poly(aryl sulfone benzimidazole) (SO(2)PBI) and poly(vinylbenzyl chloride) (PVBCl) as electrolytes for high-temperature proton-exchange-membrane fuel cells. The cross-linking imparted organo insolubility and chemical stability against radical attack to the otherwise flexible SO(2)PBI membranes. Steady phosphoric acid doping of the cross-linked membranes was achieved at elevated temperatures with little swelling. The acid-doped membranes exhibited increased mechanical strength compared to both pristine SO(2)PBI and poly[2,2'-(m-phenylene)-5,5'-bibenzimidazole] (mPBI). The superior characteristics of the cross-linked SO(2)PBI membranes allowed higher acid doping levels and, therefore, higher proton conductivity. Fuel-cell tests with the cross-linked membranes demonstrated a high open circuit voltage and improved power performance and durability.
Sigma-Aldrich
N,N-Dimethylacetamide, suitable for peptide synthesis, ā‰„99.8% (GC)
Sigma-Aldrich
Benzimidazole, 98%
Sigma-Aldrich
N,N-Dimethylacetamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlusĀ®, ā‰„99%
Supelco
N,N-Dimethylacetamide, analytical standard
Sigma-Aldrich
N,N-Dimethylacetamide, spectrophotometric grade, ā‰„99%
Sigma-Aldrich
N,N-Dimethylacetamide, puriss. p.a., ā‰„99.5% (GC)
Sigma-Aldrich
N,N-Dimethylacetamide, suitable for HPLC, ā‰„99.9%
Sigma-Aldrich
N,N-Dimethylacetamide, ReagentPlusĀ®, 99%