MilliporeSigma
Ni-catalyzed direct carboxylation of benzyl halides with CO2.
Journal of the American Chemical Society (2013-01-11)
Thierry León, Arkaitz Correa, Ruben Martin
ABSTRACT
A novel Ni-catalyzed carboxylation of benzyl halides with CO(2) has been developed. The described carboxylation reaction proceeds under mild conditions (atmospheric CO(2) pressure) at room temperature. Unlike other routes for similar means, our method does not require well-defined and sensitive organometallic reagents and thus is a user-friendly and operationally simple protocol for assembling phenylacetic acids.
Sigma-Aldrich
Benzyl chloride, ReagentPlus®, 99%, contains ≤1% propylene oxide as stabilizer
Sigma-Aldrich
Benzyl bromide, reagent grade, 98%