MilliporeSigma
  • Cyanide antidotes for mass casualties: water-soluble salts of the dithiane (sulfanegen) from 3-mercaptopyruvate for intramuscular administration.
Cyanide antidotes for mass casualties: water-soluble salts of the dithiane (sulfanegen) from 3-mercaptopyruvate for intramuscular administration.
Journal of medicinal chemistry (2013-01-11)
Steven E Patterson, Alexandre R Monteil, Jonathan F Cohen, Daune L Crankshaw, Robert Vince, Herbert T Nagasawa
ABSTRACT
Current cyanide antidotes are administered by IV infusion, which is suboptimal for mass casualties. Therefore, in a cyanide disaster, intramuscular (IM) injectable antidotes would be more appropriate. We report the discovery of the highly water-soluble sulfanegen triethanolamine as a promising lead for development as an IM injectable cyanide antidote.