MilliporeSigma
  • A simple self-assembly route to single crystalline SnO2 nanorod growth by oriented attachment for dye sensitized solar cells.
A simple self-assembly route to single crystalline SnO2 nanorod growth by oriented attachment for dye sensitized solar cells.
Nanoscale (2013-01-10)
Hui Song, Kwang-Ho Lee, Huisu Jeong, Soong Ho Um, Gil-Sang Han, Hyun Suk Jung, Gun Young Jung
ABSTRACT
One-dimensional (1-D) SnO(2) nanorods (NRs) with a rutile structure are grown on various substrates regardless of the lattice-mismatch by using a new nutrient solution based on tin oxalate, which generated supersaturated Sn(2+) sources. These affluent sources are appropriate for producing a large number of SnO(2) nanoparticles, sufficient for stacking on a substrate surface by gravity, which then acts as a seed layer for subsequent nanorod growth. Single crystalline nanorods are grown along the [001] direction by the oriented attachment phenomenon in which the attached nanoparticles were rearranged to reduce the overall surface energy through sharing thermodynamically unstable crystal (001) planes. Furthermore, the grown SnO(2) NRs are covered with a TiO(2) particulate film and utilized as a photoanode in DSSCs. The power conversion efficiency is 8.61%, enhanced by 14.2% compared to the photoanode with only a TiO(2) particulate film.
Sigma-Aldrich
Tin(IV) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(IV) oxide, −325 mesh, 99.9% trace metals basis
Sigma-Aldrich
Tin(IV) oxide, nanopowder, ≤100 nm avg. part. size