MilliporeSigma
  • A randomized, double-blind, placebo-controlled, multicenter study evaluating the efficacy, safety, and tolerability of extended-release metadoxine in adults with attention-deficit/hyperactivity disorder.
A randomized, double-blind, placebo-controlled, multicenter study evaluating the efficacy, safety, and tolerability of extended-release metadoxine in adults with attention-deficit/hyperactivity disorder.
The Journal of clinical psychiatry (2013-01-08)
Iris Manor, Rachel Ben-Hayun, Judith Aharon-Peretz, Dana Salomy, Abraham Weizman, Yaron Daniely, Dalia Megiddo, Jeffrey H Newcorn, Joseph Biederman, Lenard A Adler
ABSTRACT
To evaluate the efficacy, safety, and tolerability of an oral extended-release (ER) formulation of the nonstimulant metadoxine in the treatment of adult attention-deficit/hyperactivity disorder (ADHD). This was a 1:1 randomized, double-blind, placebo-controlled, parallel-design, phase 2 study of metadoxine ER 1,400 mg/d treatment for 6 weeks, following a 2-week baseline/screening period, involving 120 adults with DSM-IV-defined ADHD. A follow-up assessment occurred 2 weeks after the trial was completed. Efficacy measures included changes in Conners' Adult ADHD Rating Scale-Investigator Rated (CAARS-INV) total ADHD symptoms score with adult ADHD prompts (primary measure), response rates (≥ 25% or 40% improvement in CAARS-INV total ADHD symptom score), Test of Variables of Attention (TOVA) performance, and Adult ADHD Quality of Life (AAQoL) total score. The study was conducted from March 15, 2011, to August 21, 2011. Intent-to-treat analysis revealed that subjects receiving metadoxine ER showed statistically significant improvement in CAARS-INV total ADHD symptoms score (P = .02), higher rate of response (≥ 25% [P = .03] or ≥ 40% [P = .04] improvement) on the CAARS-INV total ADHD symptoms score, and improvement in TOVA score (P = .02) and AAQoL score (P = .01) compared with the placebo group. Improvements in ADHD symptoms (scored by CAARS-INV) were significantly different in subjects treated with metadoxine ER versus placebo as early as 2 weeks following treatment initiation. Metadoxine ER was generally well tolerated, with nausea (17% [10/58] vs 0% [0/59]), fatigue (31% [18/58] vs 27% [16/59]), and headaches (29% [17/58] vs 39% [23/59]) being the most frequently reported adverse effects for the metadoxine ER and placebo groups, respectively. Findings suggest that metadoxine ER is a well-tolerated and effective treatment for adults with ADHD. ClinicalTrials.gov identifier: NCT01243242.
Product Description
Supelco
Pyridoxine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Pyridoxine Hydrochloride (B6), analytical standard
Sigma-Aldrich
Pyridoxine hydrochloride, meets USP testing specifications
Sigma-Aldrich
Pyridoxine hydrochloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Pyridoxine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
L-Pyroglutamic acid, BioXtra
Sigma-Aldrich
L-Pyroglutamic acid, ≥99.0% (T)
Pyridoxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pyridoxine, ≥98%