MilliporeSigma
  • Selective serotonin reuptake inhibitors (SSRIs) inhibit insulin secretion and action in pancreatic β cells.
Selective serotonin reuptake inhibitors (SSRIs) inhibit insulin secretion and action in pancreatic β cells.
The Journal of biological chemistry (2013-01-01)
Roi Isaac, Sigalit Boura-Halfon, Diana Gurevitch, Alla Shainskaya, Yechiel Levkovitz, Yehiel Zick
ABSTRACT
Selective serotonin reuptake inhibitors (SSRIs) are antidepressants used for the treatment of mood and anxiety disorders. Here, we demonstrate that incubation (2 h) of murine islets or Min6 β cell line with the SSRIs paroxetine, fluoxetine, or sertraline inhibited insulin-induced Tyr phosphorylation of insulin receptor substrate (IRS)-2 protein and the activation of its downstream targets Akt and the ribosomal protein S6 kinase-1 (S6K1). Inhibition was dose-dependent with half-maximal effects at ∼15-20 μM. It correlated with a rapid dephosphorylation and activation of the IRS kinase GSK3β. Introduction of GSK3β siRNAs eliminated the inhibitory effects of the SSRIs. Inhibition of IRS-2 action by 30 μM SSRI was associated with a marked inhibition of glucose-stimulated insulin secretion from murine and human pancreatic islets. Secretion induced by basic secretagogues (KCl and Arg) was not affected by these drugs. Prolonged treatment (16 h) of Min6 cells with sertraline resulted in the induction of inducible nitric oxide synthase; activation of endoplasmic reticulum stress, and the initiation of the unfolded protein response, manifested by enhanced transcription of ATF4 and C/EBP homologous protein. This triggered an apoptotic process, manifested by enhanced caspase 3/7 activity, which resulted in β cell death. These findings implicate SSRIs as inhibitors of IRS protein function and insulin action through the activation of GSK3β. They further suggest that SSRIs inhibit insulin secretion; induce the unfolded protein response; activate an apoptotic process, and trigger β cell death. Given that SSRIs promote insulin resistance while inhibiting insulin secretion, these drugs might accelerate the transition from an insulin-resistant state to overt diabetes.
Product Description
Paroxetine for system suitability, European Pharmacopoeia (EP) Reference Standard
Paroxetine hydrochloride hemihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Paroxetine hydrochloride hemihydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Paroxetine maleate salt, ≥98% (HPLC), solid
Supelco
Paroxetine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®