MilliporeSigma
  • Enhanced ethanol production from wheat straw by integrated storage and pre-treatment (ISP).
Enhanced ethanol production from wheat straw by integrated storage and pre-treatment (ISP).
Enzyme and microbial technology (2013-01-01)
Volkmar Passoth, Muhammad Rizwan Tabassum, Harikrishnan A S Nair, Matilda Olstorpe, Ievgeniia Tiukova, Jerry Ståhlberg
ABSTRACT
Integrated storage and pre-treatment (ISP) combines biopreservation of moist material under airtight conditions and pre-treatment. Moist wheat straw was inoculated with the biocontrol yeast Wickerhamomyces anomalus, the xylan degrading yeast Scheffersomyces stipitis or a co-culture of both. The samples and non-inoculated controls were stored at 4 or 15 °C. The non-inoculated controls were heavily contaminated with moulds, in contrast to the samples inoculated with W. anomalus or S. stipitis. These two yeasts were able to grow on wheat straw as sole source of nutrients. When ethanol was produced from moist wheat straw stored for four weeks at 4 °C with S. stipitis, an up to 40% enhanced yield (final yield 0.15 g ethanol per g straw dry weight) was obtained compared to a dry sample (0.107 g/g). In all other moist samples, stored for four weeks at 4 °C or 15 °C, 6-35% higher yields were obtained. Thus, energy efficient bio-preservation can improve the pre-treatment efficiency for lignocellulose biomass, which is a critical bottleneck in its conversion to biofuels.
Product Description
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC