MilliporeSigma
  • Cardiac stress test as a risk-stratification tool for posttransplant cardiac outcomes in diabetic kidney transplant recipients.
Cardiac stress test as a risk-stratification tool for posttransplant cardiac outcomes in diabetic kidney transplant recipients.
Transplantation (2012-12-25)
Neeraj Singh, Samir Parikh, Udayan Bhatt, Jon Vonvisger, Uday Nori, Ayesha Hasan, Srinivas Samavedi, Kenneth Andreoni, Mitchell Henry, Ronald Pelletier, Amer Rajab, Elmahdi Elkhammas, Todd Pesavento
ABSTRACT
The utility of cardiac stress testing as a risk-stratification tool before kidney transplantation remains debatable owing to discordance with coronary angiography and outcome yields at different centers. We conducted a retrospective study of 273 diabetic kidney transplant recipients from 2006 to 2010. By protocol, all diabetic patients underwent pharmacological radionucleotide stress test or dobutamine stress echocardiography before transplant. We compared the 1-year cardiac outcomes between those with negative stress test results and those with positive stress test results. Patients with a positive stress test result (n=67) underwent coronary angiogram, and significant coronary artery disease (≥70% coronary stenosis) was found in 35 (52.2%) patients. Of the latter, 32 (91.4%) underwent cardiac revascularization (24 underwent cardiac stenting and 8 underwent coronary artery bypass grafting). The rest (n=35) were treated medically. Within 1 year after transplant, the group with positive stress test results experienced more cardiac events (34.3% vs. 3.9%, P<0.001) including acute myocardial infarction (22.4% vs. 3.4%, P<0.001) and ventricular arrhythmias (8.9% vs. 0.05%, P=0.001), higher all-cause mortality (19.4% vs. 4.8%, P<0.001), and cardiac mortality (17.9% vs. 0.9%, P<0.001) compared with the group with negative stress test results. In this diabetic population, stress testing showed positive and negative predictive values of 34.3% and 96.1%, respectively. Pharmacological cardiac stress testing provided excellent risk stratification in diabetic kidney transplant recipients.
Product Description
Dobutamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dobutamine hydrochloride, ≥98%