MilliporeSigma
  • Vitamin B12S-promoted model rearrangement of methylmalonate to succinate is not a free radical reaction.
Vitamin B12S-promoted model rearrangement of methylmalonate to succinate is not a free radical reaction.
Proceedings of the National Academy of Sciences of the United States of America (1990-04-01)
G Y Choi, S C Choi, A Galan, B Wilk, P Dowd
ABSTRACT
To probe for free radical intermediates in the model methylmalonate to succinate rearrangements promoted by vitamin B12s, a model series with a pentenyl side chain radical trap has been devised. The control free radical, generated by tri-n-butyltin hydride treatment of bromomethyl-pentenylmalonate thioester, undergoes rapid cyclization to the six-membered ring, and, as anticipated, no succinate rearrangement product is detected. By contrast when the bromide is treated with vitamin B12s, little cyclized product is observed; the major product is the pentenyl succinate. This result demonstrates that the latter rearrangement does not follow a free radical pathway.
Sigma-Aldrich
Dimethyl malonate, 98%
Sigma-Aldrich
Dimethyl malonate, purum, ≥96.0% (GC)