MilliporeSigma
  • Effects of amphiphilic star-shaped poly(ethylene glycol) polymers with a cholic acid core on human red blood cell aggregation.
Effects of amphiphilic star-shaped poly(ethylene glycol) polymers with a cholic acid core on human red blood cell aggregation.
Journal of the mechanical behavior of biomedical materials (2012-12-25)
Florence Janvier, Julian X X Zhu, Jonathan Armstrong, Herbert J Meiselman, Guy Cloutier
ABSTRACT
Elevated red blood cell (RBC) aggregation increases low-shear blood viscosity and is closely related to several pathophysiological diseases such as atherosclerosis, thrombosis, diabetes, hypertension, cancer, and hereditary chronic hemolytic conditions. Non-ionic linear polymers such as poly(ethylene glycol) (PEG) and Pluronic F68 have shown inhibitory effects against RBC aggregation. However, hypersensitivity reactions in some individuals, attributed to a diblock component of Pluronic F68, have been reported. Therefore, we investigated the use of an amphiphilic star-shaped PEG polymer based on a cholic acid core as a substitute for Pluronics to reduce RBC aggregation. Cholic acid is a natural bile acid produced in the human liver and therefore should assure biocompatibility. Cholic acid based PEG polymers, termed CA(PEG)(4), were synthesized by anionic polymerization. Size exclusion chromatography indicated narrow mass distributions and hydrodynamic radii less than 2 nm were calculated. The effects of CA(PEG)(4) on human RBC aggregation and blood viscosity were investigated and compared to linear PEGs by light transmission aggregometry. Results showed optimal reduction of RBC aggregation for molar masses between 10 and 16 kDa of star-shaped CA(PEG)(4) polymers. Cholic acid based PEG polymers affect the rheology of erythrocytes and may find applications as alternatives to linear PEG or Pluronics to improve blood fluidity.
Product Description
Cholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%