MilliporeSigma
  • Melittin-loaded immunoliposomes against viral surface proteins, a new approach to antiviral therapy.
Melittin-loaded immunoliposomes against viral surface proteins, a new approach to antiviral therapy.
Antiviral research (2012-12-25)
Alberto Falco, Enrique Barrajón-Catalán, María P Menéndez-Gutiérrez, Julio Coll, Vicente Micol, Amparo Estepa
ABSTRACT
In this study, melittin, a well-characterized pore-forming lytic amphiphilic peptide susceptible to be vehiculized in lipid membranes, has been utilized to study their antiviral properties. For this purpose, an assay based on melittin loaded-immunoliposomes previously described by our group was adapted to antiviral purposes by means of monoclonal antibodies targeting the surface G glycoprotein of the fish viral haemorrhagic septicemia rhabdovirus (VHSV). We also studied the antiviral action of these immunoliposomes in vitro and the results showed that they are capable of inhibiting the VHSV infectivity by 95.2% via direct inactivation of the virus. Furthermore, the inhibition of the infectivity when treatments were added at different times post-infection and the analysis of the infection foci sizes suggested altogether that they also act by reducing the VHSV spread in cell culture and by killing the infected cells which express the G glycoprotein in their plasmatic membranes.
Sigma-Aldrich
Melittin, ≥97% (HPLC), synthetic
Sigma-Aldrich
Melittin from honey bee venom, ≥65% (HPLC)
Sigma-Aldrich
Melittin from honey bee venom, ≥85% (HPLC)