MilliporeSigma
  • Synthesis, in vitro antibacterial and antifungal activity of some n-acetylated and non-acetylated pyrazolines.
Synthesis, in vitro antibacterial and antifungal activity of some n-acetylated and non-acetylated pyrazolines.
Pakistan journal of pharmaceutical sciences (2012-12-25)
Muhammad Baseer, Farzana Latif Ansari, Zaman Ashraf
ABSTRACT
Fourteen new N-acetylated and non-acetylated pyrazoline derivatives were synthesized by reacting chalcones with hydrazine in the presence of absolute ethanol however reaction was carried out in the presence of glacial acetic acid to afford N-acetylated pyrazolines. The chemical structures of the synthesized pyrazolines were confirmed by FTIR, (1)HNMR, (13)CNMR and mass spectroscopic data. The pyrazolines (1-14) were screened for antibacterial activity against ten bacterial strains using seven Gram-positive and three Gram-negative bacteria and antifungal activity against Aspergillus flavus, Aspergillus niger and Aspergillus pterus. Pyrazolines (1-14) found to exhibit good to excellent antimicrobial activities compared to the levofloxacin and fluconazole used as standard drugs.
Sigma-Aldrich
Hydrazine solution, 1 M in acetonitrile
Sigma-Aldrich
Hydrazine solution, 1.0 M in THF
Sigma-Aldrich
Hydrazine hemisulfate salt, ≥98%
Sigma-Aldrich
Hydrazine solution, 35 wt. % in H2O
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Hydrazine solution, 1.0 M in ethanol
Sigma-Aldrich
Hydrazine sulfate salt, puriss. p.a., ACS reagent, ≥99.0%
Sigma-Aldrich
Hydrazine dihydrochloride, ≥98%
Sigma-Aldrich
Hydrazine sulfate salt, ACS reagent, ≥99.0%