MilliporeSigma
  • Insights into sphingolipid miscibility: separate observation of sphingomyelin and ceramide N-acyl chain melting.
Insights into sphingolipid miscibility: separate observation of sphingomyelin and ceramide N-acyl chain melting.
Biophysical journal (2012-12-25)
Sherry S W Leung, Jon V Busto, Amir Keyvanloo, Félix M Goñi, Jenifer Thewalt
ABSTRACT
Ceramide produced from sphingomyelin in the plasma membrane is purported to affect signaling through changes in the membrane's physical properties. Thermal behavior of N-palmitoyl sphingomyelin (PSM) and N-palmitoyl ceramide (PCer) mixtures in excess water has been monitored by ²H NMR spectroscopy and compared to differential scanning calorimetry (DSC) data. The alternate use of either perdeuterated or proton-based N-acyl chain PSM and PCer in our ²H NMR studies has allowed the separate observation of gel-fluid transitions in each lipid in the presence of the other one, and this in turn has provided direct information on the lipids' miscibility over a wide temperature range. The results provide further evidence of the stabilization of the PSM gel state by PCer. Moreover, overlapping NMR and DSC data reveal that the DSC-signals parallel the melting of the major component (PSM) except at intermediate (20 and 30 mol %) fractions of PCer. In such cases, the DSC endotherm reports on the presumably highly cooperative melting of PCer. Up to at least 50 mol % PCer, PSM and PCer mix ideally in the liquid crystalline phase; in the gel phase, PCer becomes incorporated into PSM:PCer membranes with no evidence of pure solid PCer.
Sigma-Aldrich
N-Palmitoyl-D-sphingosine, ≥98.0% (TLC)