MilliporeSigma
  • Topochemical polymerization of phenylacetylene macrocycles: a new strategy for the preparation of organic nanorods.
Topochemical polymerization of phenylacetylene macrocycles: a new strategy for the preparation of organic nanorods.
Journal of the American Chemical Society (2012-12-20)
Simon Rondeau-Gagné, Jules Roméo Néabo, Maude Desroches, Jérémie Larouche, Josée Brisson, Jean-François Morin
ABSTRACT
Soluble organic nanorods were prepared from phenylacetylene macrocycles using the topochemical polymerization of butadiyne moieties placed both inside and outside the macrocycles' skeletons. Macrocycles containing amide groups were self-assembled in a columnar fashion through the formation of an organogel in ethyl acetate. Upon irradiation with UV light, the Raman signals associated with butadiyne units completely vanished, indicating the creation of covalently linked nanorods.
Sigma-Aldrich
Phenylacetylene, 98%