MilliporeSigma
  • Phthalate and di-(2-ethylhexyl) adipate (DEHA) intake by German infants based on the results of a duplicate diet study and biomonitoring data (INES 2).
Phthalate and di-(2-ethylhexyl) adipate (DEHA) intake by German infants based on the results of a duplicate diet study and biomonitoring data (INES 2).
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-12-19)
Hermann Fromme, Ludwig Gruber, Ralph Schuster, Martin Schlummer, Mandy Kiranoglu, Gabriele Bolte, Wolfgang Völkel
ABSTRACT
Phthalates as well as di-(2-ethylhexyl) adipate (DEHA) are used as plasticizers in diverse applications and are of toxicological concern. The study was conducted with a study population of 25 German subjects aged between 15 and 21 months. Overall, 16 phthalates and DEHA were measured by gas chromatography-mass spectrometry in a total of 171 duplicate diet samples collected over 7 consecutive days, and 20 phthalate metabolites were analyzed in the urine samples collected over 7 consecutive days using a liquid chromatography-tandem mass spectrometry method. The median "high" daily dietary intake based on 95th percentiles was 4.66 μg/kg b.w. for di-2-ethylhexyl phthalate (DEHP), 1.03 μg/kg b.w. for di-isobutyl phthalate (DiBP), and 0.70 μg/kg b.w. for di-n-butyl phthalate (DnBP), and 1.0 μg/kg b.w. for DEHA. The "high" daily total intake from biomonitoring data was 4.9 μg/kg b.w. for DEHP, 2.2 μg/kg b.w. for DnBP, 3.9 μg/kg b.w. for DiBP, and 2.6 μg/kg b.w. for di-isononyl phthalate. The comparison of the two intake estimates indicates that the dominant intake source of DEHP was food ingestion, whereas other sources considerably contributed to the total intake of other phthalates. Using our "high" intake scenario, none of the analyzed phthalates reached the recommended tolerable daily intake levels.
Product Description
Supelco
Di-n-butyl phthalate solution, certified reference material, 5000 μg/mL in methanol
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bis(2-ethylhexyl) phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Potassium hydrogen phthalate, BioXtra, ≥99.95%
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Sigma-Aldrich
Phthalic acid dipotassium salt, 98%
Sigma-Aldrich
Potassium phthalate monobasic, ACS reagent, acidimetric standard
Supelco
Bis(2-ethylhexyl) phthalate solution, certified reference material, 2000 μg/mL in methanol
Supelco
Potassium phthalate monobasic, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Potassium phthalate monobasic, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium phthalate monobasic, puriss. p.a., ≥99.5% (T)
Supelco
Potassium phthalate monobasic, reference material for titrimetry, certified by BAM, >99.5%