MilliporeSigma
  • Matrix assisted ionization vacuum (MAIV), a new ionization method for biological materials analysis using mass spectrometry.
Matrix assisted ionization vacuum (MAIV), a new ionization method for biological materials analysis using mass spectrometry.
Molecular & cellular proteomics : MCP (2012-12-18)
Ellen D Inutan, Sarah Trimpin
ABSTRACT
The introduction of electrospray ionization (ESI) and matrix-assisted laser desorption/ionization (MALDI) for the mass spectrometric analysis of peptides and proteins had a dramatic impact on biological science. We now report that a wide variety of compounds, including peptides, proteins, and protein complexes, are transported directly from a solid-state small molecule matrix to gas-phase ions when placed into the vacuum of a mass spectrometer without the use of high voltage, a laser, or added heat. This ionization process produces ions having charge states similar to ESI, making the method applicable for high performance mass spectrometers designed for atmospheric pressure ionization. We demonstrate highly sensitive ionization using intermediate pressure MALDI and modified ESI sources. This matrix and vacuum assisted soft ionization method is suitable for the direct surface analysis of biological materials, including tissue, via mass spectrometry.
Sigma-Aldrich
3-Nitrobenzonitrile, 98%
Pricing and availability is not currently available.