MilliporeSigma
Conformational analysis of flephedrone using quantum mechanical models.
Journal of molecular modeling (2012-12-15)
Wojciech Kolodziejczyk, Jerzy Jodkowski, Tiffani M Holmes, Glake A Hill
ABSTRACT
Flephedrone is an analogue of cathinone - chemically similar to ephedrine, cathine and other amphetamines. Conformations of all isomers of flephedrone have been studied at the quantum-chemical level. Calculations have been performed using DFT and MP2 methods with two basis sets - 6-31G and 6-31G(d,p). Results show that there are low energy conformers for the ortho, meta, and para isomers that are connected by way of low-barrier transition states. Boltzmann distribution of population predicts the highest population for the 1-meta conformer with a 10% increase in solution. The molecular electrostatic potential surface data for each molecule has been calculated revealing likely reaction sites.
Product Description
Supelco
R(+)-Cathinone hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®