MilliporeSigma
  • SAXS investigations of the morphology of swift heavy ion tracks in α-quartz.
SAXS investigations of the morphology of swift heavy ion tracks in α-quartz.
Journal of physics. Condensed matter : an Institute of Physics journal (2012-12-15)
B Afra, M D Rodriguez, C Trautmann, O H Pakarinen, F Djurabekova, K Nordlund, T Bierschenk, R Giulian, M C Ridgway, G Rizza, N Kirby, M Toulemonde, P Kluth
ABSTRACT
The morphology of swift heavy ion tracks in crystalline α-quartz was investigated using small angle x-ray scattering (SAXS), molecular dynamics (MD) simulations and transmission electron microscopy. Tracks were generated by irradiation with heavy ions with energies between 27 MeV and 2.2 GeV. The analysis of the SAXS data indicates a density change of the tracks of ~2 ± 1% compared to the surrounding quartz matrix for all irradiation conditions. The track radii only show a weak dependence on the electronic energy loss at values above 17 keV nm(-1), in contrast to values previously reported from Rutherford backscattering spectrometry measurements and expectations from the inelastic thermal spike model. The MD simulations are in good agreement at low energy losses, yet predict larger radii than SAXS at high ion energies. The observed discrepancies are discussed with respect to the formation of a defective halo around an amorphous track core, the existence of high stresses and/or the possible presence of a boiling phase in quartz predicted by the inelastic thermal spike model.
Sigma-Aldrich
Silicon dioxide, ~99%, 0.5-10 μm (approx. 80% between 1-5 μm)
Sigma-Aldrich
Sand, 50-70 mesh particle size