MilliporeSigma
  • A vitamin B-12 supplement of 500 μg/d for eight weeks does not normalize urinary methylmalonic acid or other biomarkers of vitamin B-12 status in elderly people with moderately poor vitamin B-12 status.
A vitamin B-12 supplement of 500 μg/d for eight weeks does not normalize urinary methylmalonic acid or other biomarkers of vitamin B-12 status in elderly people with moderately poor vitamin B-12 status.
The Journal of nutrition (2012-12-14)
Marilyn H Hill, Janet E Flatley, Margo E Barker, Clare M Garner, Nigel J Manning, Simon E Olpin, Stuart J Moat, Jean Russell, Hilary J Powers
ABSTRACT
Plasma vitamin B-12 is the most commonly used biomarker of vitamin B-12 status, but the predictive value for low vitamin B-12 status is poor. The urinary methylmalonic acid (uMMA) concentration has potential as a functional biomarker of vitamin B-12 status, but the response to supplemental vitamin B-12 is uncertain. A study was conducted to investigate the responsiveness of uMMA to supplemental vitamin B-12 in comparison with other biomarkers of vitamin B-12 status [plasma vitamin B-12, serum holotranscobalamin (holoTC), plasma MMA] in elderly people with moderately poor vitamin B-12 status. A double-blind, placebo-controlled, randomized 8-wk intervention study was carried out using vitamin B-12 supplements (500 μg/d, 100 μg/d, and 10 μg/d cyanocobalamin) in 100 elderly people with a combined plasma vitamin B-12 <250 pmol/L and uMMA ratio (μmol MMA/mmol creatinine) >1.5. All biomarkers had a dose response to supplemental vitamin B-12. Improvements in plasma vitamin B-12 and serum holoTC were achieved at cobalamin supplements of 10 μg/d, but even 500 μg/d for 8 wk did not normalize plasma vitamin B-12 in 8% and serum holoTC in 12% of people. The response in uMMA was comparable with plasma MMA; 15-25% of people still showed evidence of metabolic deficiency after 500 μg/d cobalamin for 8 wk. There was a differential response in urinary and plasma MMA according to smoking behavior; the response was enhanced in ex-smokers compared with never-smokers. uMMA offers an alternative marker of metabolic vitamin-B12 status, obviating the need for blood sampling.
Sigma-Aldrich
Methylmalonic acid, 99%
Supelco
Methylmalonic acid solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®