MilliporeSigma
  • New ruthenium sensitizers featuring bulky ancillary ligands combined with a dual functioned coadsorbent for high efficiency dye-sensitized solar cells.
New ruthenium sensitizers featuring bulky ancillary ligands combined with a dual functioned coadsorbent for high efficiency dye-sensitized solar cells.
ACS applied materials & interfaces (2012-12-14)
Yongbo Shi, Mao Liang, Lina Wang, Hongyu Han, Lingshan You, Zhe Sun, Song Xue
ABSTRACT
Two ruthenium complexes featuring bulky ancillary ligands, XS48 and XS49, were synthesized and studied as dyes in dye-sensitized solar cells (DSCs). Both dyes exhibit higher solar-to-electrical energy conversion efficiency when compared to a commonly used N3 sensitizer under the same conditions. To examine the influence of the bulky ancillary ligands and alleviate the electron recombination in cells, we have developed a dual functioned truxene-based coadsorbent (MXD1) as an alternative candidate to chenodeoxycholic acid (CDCA). This coadsorbent not only effectively shields the back electron transfer from the TiO(2) to I(3)(-) ions but also enhances the light harvesting ability in the short wavelength regions. The photovoltaic performance of XS48-sensitized DSC was independent of the coadsorbents, while XS49 with large bulky ancillary ligand presented better performance when coadsorbent was employed. Interestingly, the simultaneous adsorption-to-sequential adsorption of XS48/49 and MXD1 has caused a notably improved photovoltage, which can be primarily ascribed to the enhanced dye adsorption and retardation of charge recombination. These results not only provide a new vision on how ancillary ligands affect the performance of ruthenium complexes but also open up a new way to achieve further efficiency enhancement of ruthenium complexes.
Sigma-Aldrich
Chenodeoxycholic acid
Chenodeoxycholic acid, European Pharmacopoeia (EP) Reference Standard