MilliporeSigma
  • Electrochemical gas sensors based on paper-supported room-temperature ionic liquids for improved analysis of acid vapours.
Electrochemical gas sensors based on paper-supported room-temperature ionic liquids for improved analysis of acid vapours.
Analytical and bioanalytical chemistry (2012-12-13)
Rosanna Toniolo, Nicolò Dossi, Andrea Pizzariello, Alice Casagrande, Gino Bontempelli
ABSTRACT
A prototype of a fast-response task-specific amperometric gas sensor based on paper-supported room-temperature ionic liquids (RTILs) is proposed here for improved analysis of volatile acid species. It consists of a small filter paper foil soaked with a RTIL mixture containing an ionic liquid whose anion (acetate) displays a basic character, upon which three electrodes are screen printed by carbon ink profiting from a suitable mask. It takes advantage of the high electrical conductivity and negligible vapour pressure of RTILs and of their easy immobilization into a porous and inexpensive supporting material such as paper. The performance of this device, used as a wall-jet amperometric detector for flow injection analyses of headspace samples in equilibrium with aqueous solutions at controlled concentrations, was evaluated for phenol and 1-butanethiol vapours which were adopted as model acid gaseous analytes. The results obtained showed that the quite high potentials required for the detection of these analytes are lowered significantly, thanks to the addition of the basic acetate RTIL. In such a way, overlap with the medium discharge is avoided, and the possible adverse effect of interfering species is minimised. The sensor performance was quite satisfactory (detection limits, ca. 0.3 μM; dynamic range, ca. 1-200 μM, both referred to solution concentrations; correlation coefficients in the range 0.993-0.997; repeatability, ± 6% RSD; long-term stability, 9%); thus suggesting the possible use of this device for manifold applications.
Sigma-Aldrich
1-Butanethiol, 99%
Sigma-Aldrich
1-Butanethiol, ≥98%