MilliporeSigma
  • A tone analyzer based on a piezoelectric polymer and organic thin film transistors.
A tone analyzer based on a piezoelectric polymer and organic thin film transistors.
The Journal of the Acoustical Society of America (2012-12-13)
Yu-Jen Hsu, Ioannis Kymissis
ABSTRACT
A tone analyzer is demonstrated using a distributed resonator architecture on a tensioned piezoelectric polyvinyledene diuoride (PVDF) sheet. This sheet is used as both the resonator and detection element. Two architectures are demonstrated; one uses distributed, directly addressed elements as a proof of concept, and the other integrates organic thin film transistor-based transimpedance amplifiers directly with the PVDF to convert the piezoelectric charge signal into a current signal. The PVDF sheet material is instrumented along its length, and the amplitude response at 15 sites is recorded and analyzed as a function of the frequency of excitation. The determination of the dominant component of an incoming tone is demonstrated using linear system decomposition of the time-averaged response of the sheet and is performed without any time domain analysis. This design allows for the determination of the spectral composition of a sound using the mechanical signal processing provided by the amplitude response and eliminates the need for time-domain downstream signal processing of the incoming signal.
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~530,000, pellets
Sigma-Aldrich
Poly(vinylidene fluoride), powder
Sigma-Aldrich
Poly(vinylidene fluoride), beads or pellets
Sigma-Aldrich
Poly(vinylidene fluoride), average Mw ~275,000 by GPC, average Mn ~107,000, pellets