MilliporeSigma
  • Thermoreversible nasal in situ gel of venlafaxine hydrochloride: formulation, characterization, and pharmacodynamic evaluation.
Thermoreversible nasal in situ gel of venlafaxine hydrochloride: formulation, characterization, and pharmacodynamic evaluation.
AAPS PharmSciTech (2012-12-12)
Mandar J Bhandwalkar, Amelia M Avachat
ABSTRACT
In order to improve the bioavailability of the antidepressant drug, venlafaxine hydrochloride, in situ mucoadhesive thermoreversible gel, was formulated using Lutrol F127 (18%) as a thermo gelling polymer. Mucoadhesion was modulated by trying carbopol 934, PVP K30, HPMC K4M, sodium alginate, tamarind seed gum, and carrageenan as mucoadhesive polymers. Results revealed that as the concentration of mucoadhesive polymer increased the mucoadhesive strength increased but gelation temperature decreased. Formulation was optimized on the basis of clarity, pH, gelation temperature, mucoadhesive strength, gel strength, viscosity, drug content, diffusion through sheep nasal mucosa, histopathological evaluation of mucosa, and pharmacodynamic study in rats. Final formulation T5 containing 18% Lutrol F127 and 0.3% PVP K30 was considered as an optimized formulation. T5 released 97.86±0.073% drug in 150 min with a flux of 0.1545 mg cm(-2) min(-1) and gelation temperature 31.17±0.30°C. Histopathological evaluation of nasal mucosa revealed that T5 formulation was safe for nasal administration as it caused no damage to nasal epithelium. From the results of pharmacodynamic study, mainly forced swim test (FST), it was concluded that venlafaxine hydrochloride was more effective as an antidepressant by nasal route as in situ gel nasal drops in comparison to oral administration of equivalent dose.
Sigma-Aldrich
Venlafaxine hydrochloride, ≥98% (HPLC), powder
Pricing and availability is not currently available.
Supelco
Venlafaxine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Venlafaxine for system suitability, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Venlafaxine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.