MilliporeSigma
  • Phytochemical indicaxanthin suppresses 7-ketocholesterol-induced THP-1 cell apoptosis by preventing cytosolic Ca(2+) increase and oxidative stress.
Phytochemical indicaxanthin suppresses 7-ketocholesterol-induced THP-1 cell apoptosis by preventing cytosolic Ca(2+) increase and oxidative stress.
The British journal of nutrition (2012-12-12)
Luisa Tesoriere, Alessandro Attanzio, Mario Allegra, Carla Gentile, Maria A Livrea
ABSTRACT
7-Ketocholesterol (7-KC)-induced apoptosis of macrophages is considered a key event in the development of human atheromas. In the present study, the effect of indicaxanthin (Ind), a bioactive pigment from cactus pear fruit, on 7-KC-induced apoptosis of human monocyte/macrophage THP-1 cells was investigated. A pathophysiological condition was simulated by using amounts of 7-KC that can be reached in human atheromatous plaque. Ind was assayed within a micromolar concentration range, consistent with its plasma level after dietary supplementation with cactus pear fruit. Pro-apoptotic effects of 7-KC were assessed by cell cycle arrest, exposure of phosphatidylserine at the plasma membrane, variation of nuclear morphology, decrease of mitochondrial trans-membrane potential, activation of Bcl-2 antagonist of cell death and poly(ADP-ribose) polymerase-1 cleavage. Kinetic measurements within 24 h showed early formation of intracellular reactive oxygen species over basal levels, preceding NADPH oxidase-4 (NOX-4) over-expression and elevation of cytosolic Ca²⁺, with progressive depletion of total thiols. 7-KC-dependent activation of the redox-sensitive NF-κB was observed. Co-incubation of 2·5 μm of Ind completely prevented 7-KC-induced pro-apoptotic events. The effects of Ind may be ascribed to inhibition of NOX-4 basal activity and over-expression, inhibition of NF-κB activation, maintaining cell redox balance and Ca homeostasis, with prevention of mitochondrial damage and consequently apoptosis. The findings suggest that Ind, a highly bioavailable dietary phytochemical, may exert protective effects against atherogenetic toxicity of 7-KC at a concentration of nutritional interest.
Sigma-Aldrich
5-Cholesten-3β-ol-7-one, ≥90%