MilliporeSigma
  • The natural compounds piperovatine and piperlonguminine induce autophagic cell death on Trypanosoma cruzi.
The natural compounds piperovatine and piperlonguminine induce autophagic cell death on Trypanosoma cruzi.
Acta tropica (2012-12-12)
Phercyles Veiga-Santos, Vânia Cristina Desoti, Nathielle Miranda, Tânia Ueda-Nakamura, Benedito Prado Dias-Filho, Sueli Oliveira Silva, Diogenes Aparício Garcia Cortez, João Carlos Palazzo de Mello, Celso Vataru Nakamura
ABSTRACT
The currently available treatments for Chagas disease show limited therapeutic potential and are associated with serious side effects. Our group has been attempting to find alternative drugs isolated from natural products as a potential source of pharmacological agents against Trypanosoma cruzi. Here, we demonstrate the antitrypanosomal activity of the amides piperovatine and piperlonguminine isolated from Piper ovatum against epimastigotes and intracellular amastigotes. We also investigated the mechanisms of action of these compounds on extracellular amastigote and epimastigote forms of T. cruzi. These amides showed low toxicity to LLCMK(2) mammalian cells. By using transmission and scanning electron microscopy, we observed that the compounds caused severe alterations in T. cruzi. These alterations were mainly located in plasma membrane and mitochondria. Furthermore, the study of treated parasites labeled with Rh123, PI and MDC corroborate with our TEM data. These mitochondrial dysfunctions induced by the amides might trigger biochemical alterations that lead to cell death. Altogether, our data evidence a possible autophagic process.
Sigma-Aldrich
Sorbic acid, 99.0-101.0% anhydrous basis
Sorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sorbic acid, tested according to Ph. Eur.