MilliporeSigma
  • Persistent hyperalgesia in the cisplatin-treated mouse as defined by threshold measures, the conditioned place preference paradigm, and changes in dorsal root ganglia activated transcription factor 3: the effects of gabapentin, ketorolac, and etanercept.
Persistent hyperalgesia in the cisplatin-treated mouse as defined by threshold measures, the conditioned place preference paradigm, and changes in dorsal root ganglia activated transcription factor 3: the effects of gabapentin, ketorolac, and etanercept.
Anesthesia and analgesia (2012-12-12)
Hue Jung Park, Jennifer A Stokes, Elaine Pirie, James Skahen, Yuri Shtaerman, Tony L Yaksh
ABSTRACT
Painful neuropathy is a dose-limiting side effect in cancer chemotherapy. To characterize this phenomenon, we examined pain behavior and analgesic actions in a mouse model of cisplatin polyneuropathy. Male C57BL/6 mice received intraperitoneal cisplatin or saline (2.3 mg/kg/d) every other day 6 times over 2 weeks for a total dose of 13.8 mg/kg. Thermal escape latencies, mechanical allodynia using von Frey hairs, and observation of behavior/morbidity and body weights were assessed. After onset of allodynia, we examined the actions of intraperitoneal gabapentin (100 mg/kg), etanercept (20 and 40 mg/kg), ketorolac (15 mg/kg), and morphine (1, 3, and 10 mg/kg). Additionally, using the conditioned place preference (CPP) paradigm, we examined the effects of gabapentin and ketorolac on the presumed pain state initiated by cisplatin. Additionally, we examined the spinal cord and dorsal root ganglia (DRG) of cisplatin-treated mice. Cisplatin, but not saline treatment, produced persistent hindpaw tactile allodynia, which persisted 46 days with no effect on thermal escape. Gabapentin and morphine, but neither etanercept nor ketorolac, produced a complete but transient (2-hour) reversal of the allodynia. Etanercept (40 mg/kg) pretreatment resulted in a delay in onset of mechanical allodynia. Using CPP, gabapentin, but not ketorolac, in cisplatin animals resulted in a significant preference for the drug-associated treatment compartment. There was no place preference in non-cisplatin-treated (nonallodynic) mice after gabapentin injection. Immunohistochemistry in cisplatin-treated mice showed no change in glial fibrillary acidic protein (astrocyte) or Iba1 (ionized calcium binding adaptor molecule 1) (microglia) activation states, but a significant increase in activated transcription factor 3 was observed in the DRG. Cisplatintreated mice display allodynia and an activation of DRG activated transcription factor 3, which is paralleled by its effects on behavior in the CPP system, wherein gabapentin, but not ketorolac, in the presence of the cisplatin polyneuropathy, is positively rewarding, confirming that this neuropathy is an aversive (painful) state that is ameliorated by gabapentin.
Sigma-Aldrich
Ketorolac tris salt, ≥99%, crystalline
Ketorolac trometamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Ketorolac Tromethamine, Pharmaceutical Secondary Standard; Certified Reference Material
Ketorolac trometamol for peak identification, European Pharmacopoeia (EP) Reference Standard