MilliporeSigma
  • Induction of systemic acquired resistance by heat shock treatment in Arabidopsis.
Induction of systemic acquired resistance by heat shock treatment in Arabidopsis.
Bioscience, biotechnology, and biochemistry (2012-12-12)
Miyuki Kusajima, Soonil Kwon, Masami Nakajima, Tatsuo Sato, Takashi Yamakawa, Katsumi Akutsu, Hideo Nakashita
ABSTRACT
Systemic acquired resistance (SAR) is a potent innate immunity system in plants and has been used in rice fields. Development of SAR, involving priming, is achieved by activation of salicylic acid (SA)-mediated pathway. To determine whether heat shock (HS) treatment can induce SAR, we analyzed the effects of HS on Arabidopsis. HS treatment induced disease resistance, expression of SAR marker genes, and SA accumulation in wild-type but not in SA-deficient sid2 and NahG plants, indicating induction of SAR. Time course analysis of the effects of HS indicated that SAR was activated transiently, differently from biological induction, with a peak at 2-3 d after HS, and that it ceased in several days. Production of reactive oxygen species was observed before SA biosynthesis, which might be a trigger for SAR activation. The data presented here suggest that HS can induce SAR, but there exist unknown regulation mechanisms for the maintenance of SAR.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Supelco
Melting point standard 121-123°C, analytical standard
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Supelco
Benzoic acid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Potassium benzoate, ReagentPlus®, 99%